missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ 90mm Glass Filter Holder Accessories

Brand:  EMD Millipore™ XX1504705

Code : 72

Additional Details : Weight : 0.59596kg

 View more versions of this product

Product Code. 10094440

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Borosilicate glass construction
Specifications

Specifications

Flask
Glass
EMD Millipore 90mm Glass Filter HolderEMD Millipore 90mm Glass Filter Holder
1L
Videos
Documentation