missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ 25mm Glass Microanalysis Vacuum Filter Holder and Support

Vacuum filter small volumes for particulate or biological contamination analysis

Brand:  EMD Millipore™ XX1002530

Code : 72

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10665961

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Funnel capacity: 15mL
  • Filtration area: 2.5cm2
  • Not autoclavable with filter in place

Specifications

Specifications

Stainless Steel Support
25mm
15mL
2.5cm2
Videos
Documentation View accessory products