Learn More
EMD Millipore™ 25mm Glass Microanalysis Vacuum Filter Accessories and Replacement Parts
Accessories for filter holders for sample preparation.
Brand:  EMD Millipore™ XX1002532
Code : 72
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10676151
  • £285.00 / Each
Estimated Shipment
16-02-2018
Add to basket
Description
Description
  • Fitting Outlet: No. 5 perforated silicone stopper mounts
  • Diameter: 25mm
  • Filtration Area: 2.5 sq. cm
Specifications
Specifications
Base, stopper and screen
Stainless Steel (screen)
25mm Glass Microanalysis Vacuum Filter
Videos
Documentation