Learn More
Tocris Bioscience™ EMD 386088 hydrochloride
Potent 5-HT6 agonist
Brand:  Tocris Bioscience™ 238250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13441114
  • £356.41 / 50mg
Estimated Shipment
09-07-2019
Description
Description
Potent 5-HT6 receptor agonist (EC50 = 1.0 nM) that displays selectivity over other 5-HT receptors (IC50 values are 7.4, 110, 180, 240, 450, 620, 660 and 3000 nM for 5-HT6, 5-HT1D, 5-HT1B, 5-HT2A, 5-HT2C, 5-HT4, 5-HT1A and 5-HT7 receptors respectively). Shows moderate affinity at 5-HT3 receptors (IC50 = 34 nM).
Specifications
Specifications
5-Chloro-2-methyl-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole hydrochloride
>99%
Observed MW: 283.2
Soluble to 100mM in DMSO
1171123-46-8
C14H15N2Cl.HCl