missing translation for 'onlineSavingsMsg'
Learn More

Embigin, PE, clone: G7.43.1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5839-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15258849

  • £193.73 / 100µg


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

Description: This G7.43.1 monoclonal antibody reacts with mouse embigin (gp70), a single transmembrane glycoprotein that is a member of the Ig superfamily similar to basigin and EMMPRIN. It contains two Ig domains thought to be involved in cellular adhesion and tissue remodeling. Expression of embigin is high before day 10 in fetal development. In the adult it is expressed on myeloid cells and T cells but not B cells. During hematopoeisis, embigin is expressed on HSCs and CLPs (common lymphoid progenitors) but absent on pro- and pre-B cells. In recent studies Pax 5 has been shown to repress embigin expression.Applications Reported: This G7.43.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This G7.43.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Embigin
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P21995
embigin homolog
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
G7.43.1
PE
Emb
Conjugated
Emb
IgG2b, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
13723
Videos

For Research Use Only.