missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Elmasonic S 150 Ultrasonic Cleaning Unit, 220-240 V

The Elmasonic S 150 Ultrasonic Cleaning Unit is specifically designed and suitable for the cleaning of DIN-medicine baskets. It provides excellent cleaning and sweep technology thanks to state-of-the-art microprocessor control.

Brand:  Fisherbrand™

Additional Details : Weight : 8.00000kg

Product Code. 15879882

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • High-performance 37 kHz sandwich transducer system
  • Cleaning tank made of cavitation-resistant stainless steel
  • User-friendly and clear operating panel, splash and waterproof
  • Turning knob for setting continued and short-period operation from 1 to 30 minutes
  • Sweep function for an optimized sound field distribution in the cleaning liquid by frequency modulation
  • Degas function for the efficient degassing of the cleaning liquid and for laboratory purposes
  • Auto degas function for automatic degassing cycles, i.e. with fresh cleaning liquids
  • Ergonomically shaped plastic handles
  • Plug-in mains supply

Specifications

Specifications

1 to 30 min.
LED
340 mm
220 to 240 V
100 mm (tank)
568 mm
Stainless Steel
14 L
300 mm
224 mm
10 kg
505 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers