missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELISA-Light™ Immunoassay System with CSPD™ and Emerald-II™ Substrate/Enhancer Solution
Ultra-sensitive detection of antigens or antibodies in multiple immunoassay formats employing an alkaline phosphatase enzyme label.
Brand: Invitrogen™ T1024
Code : NEW
Additional Details : Weight : 0.00450kg
Description
- Heterogeneous assay format increases sensitivity and dynamic range and reduces interference by test compounds in the detection portion of the assay (as compared with homogeneous assay formats)
- Chemiluminescent immunoassay detection allows greater sensitivity and improved dynamic range as compared to radioimmunoassays and immunoassays that employ colorimetric or fluorescent detection
- High sensitivity and wide dynamic range permit sensitive detection with fewer assays, because sample dilution to fit the dynamic range of the assay is unnecessary
- A variety of ready-to-use substrate formulations lets the user precisely optimize signal intensity to the exact requirements of an assay
- The kinetics of the glow-assay provides a window during which measurements may be performed, facilitating HTS applications where assay automation is used
- Chemiluminescent immunoassay detection provides versatile applications such as sandwich immunoassays, competitive immunoassays, and DNA probe capture assays
- Non-radioactive immunoassays eliminate concerns over use of radioisotopes
Extend the Limits of Detection
The ELISA-Light system offers an excellent combination of high sensitivity and throughput for assays measured in a luminometer. The ELISA-Light system significantly extends the limits of detection of most immunoassays previously performed with colorimetric and isotopic methods.
Multiple Substrate/Enhancer Formulations
The kit contains reagents optimized for chemiluminescent ELISAs employing alkaline phosphatase labels and is available with a choice of ready-to-use chemiluminescent substrate formulations containing CSPD or CDP-Star™ substrate with Sapphire-II or Emerald-II™ luminescence enhancers. Multiple substrate/enhancer formulations provide options that let the user optimize specific assays or luminometers. The specific formulation that will provide the highest sensitivity for a specific assay can vary. Therefore, an ELISA sampler kit containing all four substrate/enhancer formulations is available to facilitate assay development.
Specifications
- Detection Method: Chemiluminescent
- For Use With (Equipment): Microplate Reader
- Label or Dye: AP Alkaline Phosphatase
Specifications
Immunoassay System | |
AP | |
ELISA-Light™ | |
This kit contains enough reagent for 1000 assays. Store at 4°C. |
Chemiluminescent | |
Microplate Reader | |
Alkaline Phosphatase |
For Research Use Only. Not for use in diagnostic procedures.