Learn More
GVS Elipse Mask with P3 Filters for Dust
This mask is recommended for dust and fumes containing substances. Extremely lightweight and compact to give the wearer as much comfort as possible. It offers a full range of vision without interfering with other eye or ear protection.
Brand: GVS SPR299
This item is not returnable.
View return policy
Description
This mask is recommended for dust and fumes containing substances such as micro-organisms, marble, gypsum, titanium oxide, soapstone, rock wool, wood, detergents, textile fibers, spices, salt, animal feeds, etc..
- Protects against dust that can cause lung disease. In particular, protects against coal, silica, cotton, iron ore, graphite, kaolin, zinc, aluminium dusts.
- Protects against harmful dusts such as asbestos, bauxite, coal, silica, iron, and against toxic dusts such as manganese, lead and chromium.
- Extremely lightweight and compact to give the wearer as much comfort as possible.
- Offers a full range of vision without interfering with other eye or ear protection which users are required to wear.
- Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
- Filters last up to 5 years and can be easily replaced.
- Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
- Certifications: Mask Body EN140: 1998.
- P3 RD Filters: EN143: 2000+A1: 2006.
- CE 2797
Specifications
Blue | |
Reusable |