missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mini-gel Gel Electrophoresis Unit, Battery Operated

Allows direct gel casting in tank without additional accessories.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

Product Code. 12385448

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: Horizontal mini-gel unit with built-in casting tray (2 x 1mm thick), eight sample combs, two non-rechargeable PP3 batterieseta

Description

Description

  • Complete system
  • Utilizes low voltage
  • Integrated safety lid to ensure gel remains inaccessible during use
  • UV transparent acrylic - for direct visualisation of gel within tank
  • Ultra-compact tank, minimises buffer volume required to cover gel
  • Small size to allow safe, easy transportation from bench to transilluminator
  • Built-in plastic casting gates allow direct gel casting in tank without additional costly accessories
  • Compact size: 170mm x 110mm x 35mm (D x W x H)
  • Gel dimensions: 75mm x 60mm (D x W)
  • Maximum number of samples: 32
  • Built-in gel casting tray
Specifications

Specifications

Gel Electrophoresis Unit
32
50 mL
(W x L): 6 x 7.5 cm
18 V
Videos
Documents
View accessory products