missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Electrode Cables

Brand:  Mettler Toledo™ 59902434

Code : 59

Additional Details : Weight : 0.03000kg

 View more versions of this product

Product Code. 11947971

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

RedCap S7, 4mm Banana
1m
Electrode Cable
Videos