Learn More
Scientific Industries SI™ Elastic Bands
Fastens various sample vessels to the Recessed Platform. Scientific Industries SI™ Elastic Bands can accommodate standard size microplates and vessels up to 500mL.
Brand: Scientific Industries SI™ 568-0001-00
Additional Details : Weight : 0.62000kg
Description
- Securely fastens various sample vessels to the Recessed Platform (15547325)
- Included in the Large Sample Set (Model H302, 15527325)
- Accommodates microplates and vessels up to 500ml
- For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
Specifications
Vortex-Genie 2 family of mixers, Vortex-Genie Pulse | |
Recessed Platform (15547325) |