missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Elastase, porcine pancreas aqueous suspension

Brand:  Alfa Aesar™ J61753.MB

UNSPSC : 12352204

Code : NEW

Additional Details : CAS Number : 39445-21-1 Weight : 0.00025kg

 View more versions of this product

Product Code. 15474279

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • A serine protease that acts on elastin
  • Suitable for the isolation of Type II lung cells
Specifications

Specifications

Suspension
A serine protease that acts on elastin. Suitable for the isolation of Type II lung cells
25 kd
2° to 8°C
E.C. 3.4.21.36; Pancreatopeptidase E
Off-white to Pale Brown
Elastase
25mg
39445-21-1
Documentation

Technical Documents