missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Elastase, porcine pancreas

A serine protease that acts on elastin

Brand:  Alfa Aesar™ J61874.MC

Code : NEW

Additional Details : CAS Number : 39445-21-1 Weight : 0.00010kg

 View more versions of this product

Product Code. 15484359

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Lyophilized Powder
A serine protease that acts on elastin
25 kd
2° to 8°C
E.C. 3.4.21.36; Pancreatopeptidase E
White
Elastase
100mg
39445-21-1
Soluble in water or Tris buffer up to 2.5mg/ml; Avoid high salt concentrations (ie >50mM) if salt is necessary in the buffer; Unstable in solution at pH <3.5
Videos
SDS