Learn More
Thermo Scientific™ Elastase Inhibitor II
A potent irreversible inhibitor of human neutrophil elastase
Brand: Thermo Scientific™ J65641.MA
Code : NEW
Additional Details : Weight : 0.00010kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
10 mg | |
-30°C to -10°C | |
C20H31ClN4O7 | |
HNE Inhibitor; MeOSuc-AAPA-CMK | |
Soluble in methanol or DMSO | |
Solid |
White | |
A potent irreversible inhibitor of human neutrophil elastase | |
474.90g/mol | |
Moisture Sensitive | |
Signal Transduction Reagent |