missing translation for 'onlineSavingsMsg'
Learn More

UVP™ EL Series UV Lamp

Brand:  UVP™ 95025202

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11773524

  • £454.00 / Each


Estimated Shipment
21-02-2018
Add to basket

Specifications

Specifications

376 x 96mm
White
8w
96mm
376mm
UVLMS-38
365 - 302 - 254nm
EL Series UV Lamp
Videos