Learn More
Eppendorf™ Ejector Sleeves
Extract liquids from long vessels with these ejector sleeves.
Brand:  Eppendorf™ 3110860004
Code : Z3
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11801854
  • £6.16 / Each
Estimated Shipment
21-06-2018
Add to basket
Specifications
Specifications
0.1 to 2.5uL, 0.5 to 10uL, 2 to 20uL, 10 to 100uL Pipet
1Each
Ejector Sleeve
Videos