Learn More
EIT hydrobromide, Tocris Bioscience™
Selective iNOS inhibitor, acts arginine binding site
Brand:  Tocris (Bio-Techne) 087350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13428093
  • £311.64 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Potent, selective and reversible inhibitor of isoform II NO synthase (IC 50 = 13 nM; approximately 20- and 30-fold selective over isoforms I and III respectively).
Specifications
Chemical Identifiers
1071-37-0
185.083
s-ethylisothiourea hydrobromide, 2-ethyl-2-thiopseudourea hydrobromide, ethiron, ethiron bromide, s-ethylthiuronium bromide, s-ethylisothiouronium hydrobromide, bromide s-ethylisothiuronium, s-ethylisothiuronium bromide, usaf el-18, isothuron hydrobromide
ethyl carbamimidothioate;hydrobromide
C3H9BrN2S
SWXXKWPYNMZFTE-UHFFFAOYSA-N
200213
CCSC(=N)N.Br
Specifications
50mg
1071-37-0
s-ethylisothiourea hydrobromide, 2-ethyl-2-thiopseudourea hydrobromide, ethiron, ethiron bromide, s-ethylthiuronium bromide, s-ethylisothiouronium hydrobromide, bromide s-ethylisothiuronium, s-ethylisothiuronium bromide, usaf el-18, isothuron hydrobromide
CCSC(=N)N.Br
185.083
185.08
EIT hydrobromide
C3H9BrN2S
SWXXKWPYNMZFTE-UHFFFAOYSA-N
ethyl carbamimidothioate;hydrobromide
200213
>98%