missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ EIF2C1 Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding full length of human EIF2C1 (Met 1-Ala 857) fused to polyhistidine tag at N-terminus

Brand:  Sino Biological™ 11225H07B50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15268415

  • £370.60 / 50µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

  • Formulation: 50mM Tris, 100mM NaCl, 10% glycerol, 0.5 PMSF, 0.5mM EDTA, pH 8.0
Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
26523
≥90% by SDS-PAGE
Human
<1 EU/μg
EIF2C1
Other proteins
EIF2C1 Recombinant Human Protein
Lyophilized
99.5kd
50μg
Full Length, Recombinant
Eukaryotic Translation Initiation Factor 2C 1
Assay Standard
Insect
N-His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.