Learn More
EHF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563890
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MILEGGGVMN LNPGNNLLHQ PPAWTDSYST CNVSSGFFGG QWHEIHPQYW TKYQVWEWLQ HLLDTNQLDA NCIPFQEFDI NGEHL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.
EHF Polyclonal antibody specifically detects EHF in Human samples. It is validated for Immunohistochemistry.Specifications
EHF | |
Polyclonal | |
Unconjugated | |
EHF | |
9030625L19Rik; AU019492; EHF; Epithelium-specific Ets transcription factor 3; ESE3; ESE-3; ESE3 transcription factor; ESE3B; ESEJ; ETS domain-containing transcription factor; ETS homologous factor; hEHF; Unknown (protein for MGC:133971) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26298 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZC4 | |
EHF | |
Recombinant protein corresponding to Human EHF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only