Learn More
EHD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562116
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PDNRKLFEAE EQDLFRDIQS LPRNAALRKL NDLIKRARLA KVHAYIISSL KKE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.
EHD3 plays a role in endocytic transport.Specifications
EHD3 | |
Polyclonal | |
Unconjugated | |
EHD3 | |
EH domain containing 3; EH domain-containing protein 3; Ehd2; Ehd3; EH-domain containing 3; EH-domain containing protein 2; PAST homolog 3; PAST3; si:ch211-147c1.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30845 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZN3 | |
EHD3 | |
Recombinant protein corresponding to Human EHD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only