Learn More
EGR2 Monoclonal Antibody (erongr2), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£101.00 - £242.00
Specifications
Antigen | EGR2 |
---|---|
Clone | erongr2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15300820
|
Affymetrix eBioscience
17-6691-80 |
25 μg |
£101.00
25µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15310820
|
Affymetrix eBioscience
17-6691-82 |
100 μg |
£242.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This erongr2 monoclonal antibody recognizes mouse early growth response 2 (Egr2, Krox-20). Egr2 is a zinc finger-containing transcription factor that is important for the development of T cells and NKT cells. Egr2 is predomitly expressed in the thymus and nervous system, and it is also upregulated in T cells following TCR-crosslinking. Upregulation in all non-adherent mouse splenocytes has been observed following stimulation with phorbol 12-myristate 13-acetate (PMA) and ionomycin. Egr2 is believed to antagonize T cell activation and has also been reported to promote apoptosis. The erongr2 antibody also appears to modestly stain a subpopulation of T cells and B cells in freshly isolated spleen cells. The erongr2 monoclonal antibody is specific for Egr2 and does not crossreact to Egr3, based on immunoblot analysis. Applications Reported: This erongr2 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This erongr2 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523). This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
EGR2 Monoclonal antibody specifically detects EGR2 in Mouse samples. It is validated for Flow Cytometry.Specifications
EGR2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AT591; CMT1D; CMT4E; DKFZp686J1957; E3 SUMO-protein ligase EGR2; E3 SUMO-protein transferase ERG2; early growth response 2; early growth response protein 2; Egr2; Egr-2; FLJ1454; FLJ14547; Krox20; Krox-20; Krox-20 homolog; KROX-20, Drosophila, homolog (early growth response-2); NGF1-B; OTTHUMP00000062133; Zfp-25; Zfp-6; Zinc finger protein Krox-20 | |
Egr2 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
erongr2 | |
Flow Cytometry | |
APC | |
Rat | |
Mouse | |
P08152 | |
13654 | |
E. coli-derived protein, aa 1-319 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Egr2 |
For Research Use Only.