Learn More
Egr1 Rat anti-Human, PerCP-eFluor 710, Clone: HEGR1DS, eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 46-9851-41
Code : NEW
Additional Details : Weight : 0.23750kg
Product Code. 15569436
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
The monoclonal antibody HEGR1DS recognizes human Egr1 (Early Growth Response 1), a C2H2-type zinc-finger transcription factor broadly expressed in various types of tissues. Egr1 is induced within minutes after cell activation and it regulates expression of numerous factors involved in cell division, growth, and differentiation. It has been shown to suppress tumorigenesis by inducing apoptosis of cancer cells but it can also promote their proliferation. Egr1 is often used as an early cell activation marker of T and B lymphocytes as well as myeloid cells.
Specifications
Specifications
Egr1
HEGR1DS
PerCP-eFluor 710
Conjugated
Rat
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Monoclonal
Flow Cytometry
5μL (0.015μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Zif268, NGFI-A, KROX-24, ZENK, Znf255
IgG2a, kappa