missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EGR1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | EGR1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
EGR1 Polyclonal specifically detects EGR1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
EGR1 | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
AT225Transcription factor ETR103, early growth response 1, early growth response protein 1, EGR-1, G0S30, KROX24, KROX-24, Nerve growth factor-induced protein A, NGFIA, NGFI-ATranscription factor Zif268, TIS8, ZIF-268, Zinc finger protein Krox-24, ZNF225, ZNF225Zinc finger protein 225 | |
EGR1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
P18146 | |
1958 | |
E. coli-derived recombinant human EGR1 Gln282-Ser433 Accession # P18146 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human EGR1 in direct ELISAs and Western blots. |
For Research Use Only