missing translation for 'onlineSavingsMsg'
Learn More

EGLU, Tocris Bioscience™

Highly selective group II mGlu antagonist

Brand:  Tocris (Bio-Techne) 0971/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448223

  • £135.24 / 10mg

Estimated Shipment date
from Supplier 02-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective antagonist of presynaptically-mediated (1S,3S)-ACPD-induced depression of motoneuron excitation in neonatal rat spinal cord; presumed group II mGlu receptor antagonist. Also available as part of the Group II mGlu Receptor Tocriset™ .
Specifications

Chemical Identifiers

170984-72-2
175.184
eglu, s-alpha-ethylglutamate, 2s-2-amino-2-ethylpentanedioic acid, 2s-alpha-ethylglutamic acid, tocris-0971, 2-ethyl-l-glutamic acid, d01wkr, 2s-?-ethylglutamic acid, l-glutamic acid, 2-ethyl, s-2-amino-2-ethylpentanedioic acid
(2S)-2-amino-2-ethylpentanedioic acid
C7H13NO4
QFYBYZLHPIALCZ-ZETCQYMHSA-N
5311079
CCC(CCC(=O)O)(C(=O)O)N

Specifications

EGLU
C7H13NO4
QFYBYZLHPIALCZ-ZETCQYMHSA-N
(2S)-2-amino-2-ethylpentanedioic acid
5311079
>95%
170984-72-2
eglu, s-alpha-ethylglutamate, 2s-2-amino-2-ethylpentanedioic acid, 2s-alpha-ethylglutamic acid, tocris-0971, 2-ethyl-l-glutamic acid, d01wkr, 2s-?-ethylglutamic acid, l-glutamic acid, 2-ethyl, s-2-amino-2-ethylpentanedioic acid
CCC(CCC(=O)O)(C(=O)O)N
175.184
175.18
10mg
Videos
Documents