missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat EG-VEGF/PK1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
POA
Specifications
Antigen | EG-VEGF/PK1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
EG-VEGF/PK1 Polyclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
EG-VEGF/PK1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human, Mouse, Rat | |
EGVEGF, EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, PROK1, prokineticin 1 | |
PROK1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q8R414 | |
84432 | |
Mouse myeloma cell line NS0-derived recombinant rat EG-VEGF/PK1 Ala19-Phe105 Accession # Q8R414 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human, mouse, and rat EG-VEGF/PK1 in direct ELISAs and Western blots. In direct ELISAs, 100% cross reactivity with recombinant human EG-VEGF and recombinant mouse EG-VEGF is observed |
For Research Use Only