Learn More
Tocris Bioscience™ Efonidipine hydrochloride monoethanolate
Ca2+ channel blocker (L- and T-type)
Brand:  Tocris Bioscience™ 373350
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13453514
  • £286.65 / 50mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Selective blocker of L-type and T-type Ca2+ channels. Displays minimal inhibition of N- and P/Q-type channels and no inhibition of R-type channels. R(-) and S(+)-enantiomers display different channel selectivity; S(+)-Efonidipine blocks L-type and T-type channels whereas R(-)-Efonidipine displays selectivity for T-type channels. Exhibits antihypertensive activity.
Specifications
Specifications
Efonidipine hydrochloride monoethanolate
>99
50mg
Efonidipine hydrochloride monoethanolate
714.18