missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ EFNA3 (EphrinA3) Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding mature form of human EphrinA3, residues 1 to 213, fused topolyhistidine tag at C-terminus

Brand:  Sino Biological™ 10188H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13405786

  • £56.74 / 5µg


Estimated Shipment
09-05-2018
Add to basket

Description

Description

  • Activity is measured by its ability to compete with human EFNA3-Fc for binding to immobilized mouse EFNA6-His in functional ELISA assay
  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications

Specifications

Studying protein-protein interactions and protein structure analysis
1944
≥90% by SDS-PAGE
Human
Recombinant
Ephrin-A3
Assay Standard
Mammalian
His Tag
Lyophilized
23kd
5μg
−20° to −70°C
<1 EU/μg
EFNA3
Kinases and Inhibitors
EFNA3 (EphrinA3) Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.