Learn More
Tocris Bioscience™ Efaroxan hydrochloride
Brand:  Tocris Bioscience™ 079250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13448013
  • £536.55 / 50mg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Potent, highly selective α2 adrenoceptor antagonist and imidazoline I1 receptor ligand (pKi values are 7.87, 7.42, 5.74, 7.28 and < 5 for α 2A, α2B, α2C, I1, and I2 receptors respectively). Promotes insulin secretion, at a site distinct from I1 or I2 (the putative I3 receptor) in vitro and in vivo.
Specifications
Specifications
2-(2-Ethyl-2,3-dihydro-2-benzofuranyl)-4,5-dihydro-1H-imidazole hydrochloride
>99%
Observed MW: 252.74
Soluble to 100 mM in water
89197-00-2
C13H16N2O.HCl