missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Efaroxan hydrochloride
alpha2 antagonist and I1, I3 ligand
POA
Specifications
Assay Percent Range | >99% |
---|---|
Chemical Name or Material | 2-(2-Ethyl-2,3-dihydro-2-benzofuranyl)-4,5-dihydro-1H-imidazole hydrochloride |
CAS | 89197-00-2 |
Description
Potent, highly selective α2 adrenoceptor antagonist and imidazoline I1 receptor ligand (pKi values are 7.87, 7.42, 5.74, 7.28 and < 5 for α 2A, α2B, α2C, I1, and I2 receptors respectively). Promotes insulin secretion, at a site distinct from I1 or I2 (the putative I3 receptor) in vitro and in vivo.Specifications
>99% | |
2-(2-Ethyl-2,3-dihydro-2-benzofuranyl)-4,5-dihydro-1H-imidazole hydrochloride | |
89197-00-2 |
For Research Use Only