missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ EED Recombinant Human Protein, N-His.GST tag

Expressed from DNA sequence encoding human EED (Met 1-Arg 441) fused to N-terminal polyhistidine-tagged GST tag at N-terminus

Brand:  Sino Biological™ 11307H20B250

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238515

  • £1190.91 / 250µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 2mM GSH, 10% Glycerol, pH 8.0
Specifications

Specifications

Lyophilized
78kd
5 x 50μg
Full Length, Recombinant
Embryonic Ectoderm Development
Assay Standard
Insect
GST Tag, His Tag
8726
≥90% by SDS-PAGE
Human
<1 EU/μg
EED
Other proteins
EED Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.