missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA, Disodium Salt, Dihydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™
345.80 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
- For Molecular Biology Applications
Specifications
Packaging | Poly Bottle |
Quantity | 1 kg |
IUPAC Name | disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
Formula Weight | 372.24 |
Grade | Ultrapure Bioreagent |
Physical Form | Crystals |
Chemical Name or Material | EDTA, Disodium Salt, Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'