missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ Edge™ 40-517 Series Gray Nitrile Medium Weight Gloves
For quick and accurate handling providing comfort and breathability.
Brand: Ansell Edmont™ 40157090
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Moulds to hand ensuring improved movement and dexterity
- Porous nitrile coating helps channel oil away from glove surface for firmer grip
- Fully-coated finish
- Cut and sewn construction
- Interlock cotton liner
- Safety cuff
Specifications
Foam Nitrile | |
40-517 | |
Nitrile Foam | |
Interlock Cotton | |
Superior abrasion, cuts and tearing resistance | |
9 | |
CE Mark, CATEGORY II EN 388 : 4121, EN 1149 |
Medium Weight Gloves | |
Nitrile Foam | |
Safety cuff | |
Fully coated-palm | |
40-517 | |
144 Pairs |