Learn More
EDG4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554038
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LLLDGLGCES CNVLAVEKYF LLLAEANSLV NAAVYSCRDA EMRRTFRRLL CCACLRQSTR ESVHYTSSAQ GGASTRIMLP ENGHPLMDST L Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
EDG4 Polyclonal antibody specifically detects EDG4 in Human samples. It is validated for Immunohistochemistry.Specifications
EDG4 | |
Polyclonal | |
Unconjugated | |
LPAR2 | |
Edg4; EDG-4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 4; G protein-coupled receptor; IPA2; LPA receptor 2; LPA receptor EDG4; LPA2; LPA-2; LPA2 Receptor; LPAR2; Lysophosphatidic acid receptor 2; lysophosphatidic acid receptor EDG4; lysophosphatidic acid receptor Edg-4; Pbx4; pre-B-cell leukemia transcription factor 4; RGD1561336 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBW0 | |
LPAR2 | |
Recombinant protein corresponding to Human EDG4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only