missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECM-1/Secretory Component P85 Mouse anti-Human, Clone: 966231, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | ECM-1/Secretory Component P85 |
---|---|
Clone | 966231 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ECM-1 Ala20-Glu540 Accession # Q16610 |
Host Species | Mouse |
Primary or Secondary | Primary |
Description
ECM-1/Secretory Component P85 Monoclonal specifically detects ECM-1/Secretory Component P85 in Human samples. It is validated for Western Blot, ELISA.Specifications
ECM-1/Secretory Component P85 | |
Mouse myeloma cell line NS0-derived recombinant human ECM-1 Ala20-Glu540 Accession # Q16610 | |
Primary | |
Human | |
Unconjugated | |
1893 | |
Purified | |
Western Blot, ELISA | |
RUO |
966231 | |
Mouse | |
Monoclonal | |
IgG1 | |
ECM1, extracellular matrix protein 1, p85, Secretory Component Glycoprotein, Secretory Component P85 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO