missing translation for 'onlineSavingsMsg'
Learn More

EC 23, Tocris Bioscience™

Synthetic retinoid; induces differentiation of stem cells

Brand:  Tocris (Bio-Techne) 401110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462784

  • £155.76 / 10mg


Estimated Shipment
22-02-2019
Add to basket

Description

Description

Photostable synthetic retinoid; analog of ATRA (Cat. No. 0695). Displays similar activity to ATRA when tested on the embryonal carcinoma stem cell model TERA2.cl.SP12. Potently induces neural differentiation in human pluripotent embryonic stem cells.
Specifications

Chemical Identifiers

104561-41-3
332.443
vsdtydjdjj, unii-vsdtydjdjj, 4-2-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethynyl benzoic acid, 4-2-1,1,4,4-tetramethyltetralin-6-yl ethynyl benzoic acid, 4-2-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethynyl, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethynyl benzoic acid, 4-2-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl ethynyl benzoic acid, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-ylethynyl benzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphth-2-yl-ethynyl-benzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphth-2-yl ethynyl-benzoic acid
4-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethynyl]benzoic acid
C23H24O2
OQVLOWLEEHYBJH-UHFFFAOYSA-N
10314719
CC1(CCC(C2=C1C=CC(=C2)C#CC3=CC=C(C=C3)C(=O)O)(C)C)C

Specifications

EC 23
104561-41-3
vsdtydjdjj, unii-vsdtydjdjj, 4-2-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethynyl benzoic acid, 4-2-1,1,4,4-tetramethyltetralin-6-yl ethynyl benzoic acid, 4-2-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethynyl, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethynyl benzoic acid, 4-2-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl ethynyl benzoic acid, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-ylethynyl benzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphth-2-yl-ethynyl-benzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphth-2-yl ethynyl-benzoic acid
CC1(CCC(C2=C1C=CC(=C2)C#CC3=CC=C(C=C3)C(=O)O)(C)C)C
332.443
332.44
10mg
C23H24O2
OQVLOWLEEHYBJH-UHFFFAOYSA-N
4-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethynyl]benzoic acid
10314719
>99%
Videos