missing translation for 'onlineSavingsMsg'
Learn More

EBPC, Tocris Bioscience™

Aldose reductase inhibitor

Brand:  Tocris (Bio-Techne) 051850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437683

  • £420.32 / 50mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent aldose reductase inhibitor. Improves the cytotoxicity of anticancer reagents such as cisplatin (Cat.No. 2251) and doxorubicin (Cat.No. 2252) in HeLa cervical carcinoma cells through an increase in ERK activity.
Specifications

Chemical Identifiers

4450-98-0
261.277
ebpc, 1-benzyl-4-hydroxy-5-oxo-2,5-dihydro-1h-pyrrole-3-carboxylic acid ethyl ester, ethyl 1-benzyl-3-hydroxy-2 5h-oxopyrrole-4-carboxylate, 1-benzyl-4-ethoxy hydroxy methylidene pyrrolidine-2,3-dione, ethyl 1-benzyl-4-hydroxy-5-oxo-2,5-dihydro-1h-pyrrole-3-carboxylate, 1-ebpc, ethyl 1-benzyl-3-hydroxy-2-oxo 5h pyrrole-4-carboxylate, spectrum_001772, specplus_000694, maybridge1_001985
ethyl 1-benzyl-4-hydroxy-5-oxo-2H-pyrrole-3-carboxylate
C14H15NO4
IGYRPDIWSYGHMY-UHFFFAOYSA-N
54677973
CCOC(=O)C1=C(C(=O)N(C1)CC2=CC=CC=C2)O

Specifications

EBPC
4450-98-0
ebpc, 1-benzyl-4-hydroxy-5-oxo-2,5-dihydro-1h-pyrrole-3-carboxylic acid ethyl ester, ethyl 1-benzyl-3-hydroxy-2 5h-oxopyrrole-4-carboxylate, 1-benzyl-4-ethoxy hydroxy methylidene pyrrolidine-2,3-dione, ethyl 1-benzyl-4-hydroxy-5-oxo-2,5-dihydro-1h-pyrrole-3-carboxylate, 1-ebpc, ethyl 1-benzyl-3-hydroxy-2-oxo 5h pyrrole-4-carboxylate, spectrum_001772, specplus_000694, maybridge1_001985
CCOC(=O)C1=C(C(=O)N(C1)CC2=CC=CC=C2)O
261.277
261.28
50mg
C14H15NO4
IGYRPDIWSYGHMY-UHFFFAOYSA-N
ethyl 1-benzyl-4-hydroxy-5-oxo-2H-pyrrole-3-carboxylate
54677973
>99%
Videos
Documents