missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EASY-Spray™ HPLC Columns

Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.

Brand:  Thermo Scientific™ ES804A

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15929007

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.

The unique EASY-Spray design provides uncompromised performance and ease-of-use in nano and capillary LC-MS proteomics applications. Features include the following:

  • Precision positioned glass emitter
  • Integrated precision designed union
  • Integrated temperature control
Specifications

Specifications

Analytical Column, EASY-Spray
150 mm
2 μm
100 Å
60°C
C18
Spherical, Fully Porous, Ultrapure Silica
Reversed Phase
Reversed Phase
75 μm
11600 psi (800 bar)
2 to 8
450 m²/g
L1
Easy Spray
15%
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.