Learn More
EAF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552557
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GFSHLDRRER VLKLGESFEK QPRCAFHTVR YDFKPASIDT SSEGYLEVGE GEQVTITLPN IEGSTPPVTV FKGSKKPYLK ECILIINHDT GECRL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 89%.
EAF2 Polyclonal antibody specifically detects EAF2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
EAF2 | |
Polyclonal | |
Unconjugated | |
Eaf2 | |
AW048865; BM040; BM-040; Eaf2; Ehrlich S-II transcriptional activator factor; ELL associated factor 2; ELL-associated factor 2; Festa; FESTA-L; FESTA-S; testosterone regulated apoptosis inducer and tumor suppressor; testosterone-regulated apoptosis inducer and tumor suppressor protein; Traits; U19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55840 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CJ1 | |
Eaf2 | |
Recombinant protein corresponding to Human EAF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only