missing translation for 'onlineSavingsMsg'
Learn More

Ea52-68 peptide bound to I-Ab, FITC, clone: eBioY-Ae (YAe, Y-Ae), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5741-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228249

  • £111.93 / 100µg


Estimated Shipment date
from Supplier 01-06-2018
Add to basket

Description

Description

Description: The Y-Ae antibody reacts with a class II major histocompatibility complex (MHC) self Ea peptide (peptide 52-68) bound to I-Ab molecules. The Y-Ae antibody detects a determinant expressed on a subset of class II I-Ab molecules in strains that also express class II I-Eb. This determinant is expressed on peripheral B cells and on cells in the thymic medulla, but not thymic cortical epithelium. In addition to B cells, the Y-Ae determinant is expressed at comparable levels on other antigen presenting cells, including macrophages and dendritic cells. The antibody does not react with invariant chain-associated class II MHC complexes. The reactivity of the Y-Ae antibody is MHC-restricted. The Y-Ae antibody was generated by immunization of [B10.MBR x B10.D2]F1 recipients with B10A(5R) LPS blasts. In this strain combination, there is incompatibility for I-Ab and I-Eb molecules. The antibody reacts with 10-15% of surface I-Ab molecules in strains that also express surface I-Eb molecules (e.g., strains B10.A(5R) and B10.A(3R)). The antibody does not react with I-A molecules in strains that have a non-functional I-E (a chain) gene (e.g., strains B6 and B10), nor with mutant I-A molecules (e.g., (B6.c-H-2bm-12 x A.TFR5)F1), nor with any other strain tested. The Y-Ae antibody recognizes an Ea peptide (Ea 52-68) bound to I-Ab molecules, but not to Ea 52-68 peptide bound to I-Ak or I-Abm-12 molecules.Applications Reported: This eBioY-Ae (YAe, Y-Ae) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioY-Ae (YAe, Y-Ae) antibody has been tested flow cytometric analysis of mouse peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ea52-68 peptide bound to I-Ab
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06342
H-2Eb, H2Eb, Ia-4, Ia4, MGC163794, MGC163796
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
Y-Ae), eBioY-Ae (YAe
FITC
H2-Ab1
Conjugated
H2-Ab1
IgG2b
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
14961
Videos

For Research Use Only.