Learn More
E4CPG, Tocris Bioscience™
Group I and II mGlu antagonist
Brand:  Tocris (Bio-Techne) 100950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13448283
  • £557.13 / 50mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Novel group I/group II metabotropic glutamate receptor antagonist, more potent than (RS)-MCPG (Cat. No. 0336).
Specifications
Chemical Identifiers
170846-89-6
223.228
tocris-1009, s-2-amino-2-4-carboxyphenyl butanoic acid, 4-2s-2-amino-1-hydroxy-1-oxobutan-2-yl benzoic acid
4-[(1S)-1-amino-1-carboxypropyl]benzoic acid
C11H13NO4
AIEFWRHRHFRLFT-NSHDSACASA-N
6604793
CCC(C1=CC=C(C=C1)C(=O)O)(C(=O)O)N
Specifications
E4CPG
C11H13NO4
AIEFWRHRHFRLFT-NSHDSACASA-N
4-[(1S)-1-amino-1-carboxypropyl]benzoic acid
6604793
>99%
170846-89-6
tocris-1009, s-2-amino-2-4-carboxyphenyl butanoic acid, 4-2s-2-amino-1-hydroxy-1-oxobutan-2-yl benzoic acid
CCC(C1=CC=C(C=C1)C(=O)O)(C(=O)O)N
223.228
223.23