Learn More
Thermo Scientific™ E1-ClipTip™ Multichannel Pipettes, 12-channel, 300
Versatility and customization, automate daily 96-and 384-well microplate pipetting tasks with the sealed in security of ClipTip technology.
Brand: Thermo Scientific™ 4671080
Code : NEW
Additional Details : Weight : 2.00000kg Transport : UN number : 3481 Chem class : 9
Description
With enhanced versatility and customization, the Thermo Scientific™ E1-ClipTip™ Electronic Multichannel Pipette automates daily 96-and 384-well microplate pipetting tasks with the sealed in security of Thermo Scientific™ ClipTip™ technology. Enjoy pipetting again with optimum functionality for every application and user preference. Feel the difference.
Innovative ClipTip interlocking tip interface technology†- Tip is locked and sealed in place until ejected— no dropped tips that waste valuable samples
- Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
- Tips are attached and ejected with a light touch— eliminates the frustration of banging tips on the pipette
An ideal choice for sample transfers
- Personalized user interface and password protection ensure consistent operation among multiple users.
- Electronic operation to help you enjoy pipetting again
- Designed for optimum comfort and ease of use, the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.