missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 1.2%

System starter for fast and convenient E-Gel agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel SYBR Safe 1.2% agarose gels.

Brand:  Invitrogen™ G8351ST


Additional Details : Weight : 1.10000kg

Product Code. 15706568

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: E-Gel Power Snap Electrophoresis device,E-Gel Power Snap Camera,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.

Description

Description

The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, SYBR Safe 1.2%, provides everything you need to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.

The kit includes:

  • 18 E-Gel agarose gels with SYBR Safe, 1.2% (Cat. No. G521801)
  • E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)

Services

1 year warranty

Specifications

Specifications

• 1 electrophoresis device
• 1 camera device
• 18 E-Gel SYBR Safe 1.2% gels
• 1 vial E-Gel 1 kb Plus DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 E-Gel Go! Adapter

Store at room temperature.
Electrophoresis System Starter Kit
Room Temperature
240 V AC (Power Supply), 48 V DC (Electrophoresis Device)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.