missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dysprosium, plasma standard solution, Specpure™, Dy 1000μg/mL

Brand:  Alfa Aesar™ 013838.AD

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 50mL

 View more versions of this product

Product Code. 11311399

  • £36.20 / 50mL


Estimated Shipment date
from Supplier 20-06-2018
Add to basket

Specifications

Chemical Identifiers

Dysprosium plasma standard solution
Specpure™
UN3264
Dy 1000μg/mL
MFCD00010982

Specifications

Dysprosium plasma standard solution
Specpure™
Dy2O3 in 5% HNO3
UN3264
Dy 1000μg/mL
Liquid
MFCD00010982
50mL
Videos
SDS