missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dysprosium(III) titanium oxide, 99% (metals basis)

Brand:  Alfa Aesar™ 041649.09

Code : NEW

Additional Details : CAS Number : 68993-46-4 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11368788

  • £53.00 / 10g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Dysprosium(III) titanium oxide
450.14
Dysprosium(III) titanate
68993-46-4
MFCD00467691

Specifications

Dysprosium(III) titanium oxide
Powder
(metals basis)
MFCD00467691
10g
68993-46-4
99%
450.14
Dysprosium(III) titanate
Videos
SDS