missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysprosium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
107.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Dysprosium(III) oxide is uses in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2340°C |
Physical Form | Sintered Lump |
Quantity | 25 g |
Merck Index | 14,3482 |
Solubility Information | Insoluble in water. |
Formula Weight | 373 |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Density | 7.81 g/mL at 27°C |
Assay | (REO) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'