missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dysprosium(III) isopropoxide, 99.9% (REO), 5% w/v in toluene/isopropanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Description
Dysprosium(III) isopropoxide is used as primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown |
Linear Formula | Dy[OCH(CH3)2]3 |
Quantity | 100 mL |
UN Number | UN1993 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 339.76 |
Percent Purity | 99.9% |
Concentration or Composition (by Analyte or Components) | 5% w/v in toluene/isopropanol |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'