missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dysprosium(III) bromide, ultra dry, 99.99% (REO)

Brand:  Alfa Aesar™ 038611.03

Code : NEW

Additional Details : CAS Number : 14456-48-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11338937

  • £34.03 / 1g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

Dysprosium(III) bromide
Ultra dry
MFCD00016069
14456-48-5
402.23

Specifications

Dysprosium(III) bromide
14456-48-5
99.99%
402.23
Soluble in water
Hygroscopic
881°C
Ultra dry
Powder
(REO)
MFCD00016069
Ampule
1480°C
1g
Videos
SDS