missing translation for 'onlineSavingsMsg'
Learn More

Dynole 34-2, Tocris Bioscience™

Dynamin I inhibitor

Brand:  Tocris (Bio-Techne) 422210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13464384

  • £175.56 / 10mg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Dynamin I inhibitor (IC 50 = 1.3 μM). Inhibits receptor-mediated endocytosis (RME) (IC 50 = 5.0 μM).
Specifications

Chemical Identifiers

1128165-88-7
408.59
dynole 34-2, 2e-2-cyano-3-1-3-dimethylamino propyl indol-3-yl-n-octylprop-2-enamide, 1-3-dimethylamino propyl-n-octyl-alpha-cyano-1h-indole-3-acrylamide, 2-cyano-n-octyl-3-1-3-dimethylaminopropyl-1h-indol-3-yl-acrylamide, 2-cyano-3-1-3-dimethylamino propyl-1h-indol-3-yl-n-octyl-2-propenamide
(E)-2-cyano-3-[1-[3-(dimethylamino)propyl]indol-3-yl]-N-octylprop-2-enamide
C25H36N4O
MYIMRONQBLZJFI-DYTRJAOYSA-N
44157463
CCCCCCCCNC(=O)C(=CC1=CN(C2=CC=CC=C21)CCCN(C)C)C#N

Specifications

Dynamin Inhibitors
1128165-88-7
dynole 34-2, 2e-2-cyano-3-1-3-dimethylamino propyl indol-3-yl-n-octylprop-2-enamide, 1-3-dimethylamino propyl-n-octyl-alpha-cyano-1h-indole-3-acrylamide, 2-cyano-n-octyl-3-1-3-dimethylaminopropyl-1h-indol-3-yl-acrylamide, 2-cyano-3-1-3-dimethylamino propyl-1h-indol-3-yl-n-octyl-2-propenamide
CCCCCCCCNC(=O)C(=CC1=CN(C2=CC=CC=C21)CCCN(C)C)C#N
408.59
408.58
10mg
C25H36N4O
MYIMRONQBLZJFI-DYTRJAOYSA-N
(E)-2-cyano-3-[1-[3-(dimethylamino)propyl]indol-3-yl]-N-octylprop-2-enamide
44157463
>99%
Videos