Learn More
Thermo Scientific PCR Buffer; Long Range PCR; Thermo Scientific; Detergent-free DyNAzyme EXT Buffer; 1 x detergent-free Buffer contains: 50 mM Tris-HCl (pH 9.0 at 25 deg.C), 1.5 mM MgCl2 and 15 mM (NH4)2SO4; 4 x 1.5mL of 10 Optimized Buffer; 6mL
Brand:  Thermo Scientific F-517L
PCR Buffer; Long Range PCR; Thermo Scientific; Detergent-free DyNAzyme EXT Buffer; 1 x detergent-free Buffer contains: 50 mM Tris-HCl (pH 9.0 at 25 deg.C), 1.5 mM MgCl2 and 15 mM (NH4)2SO4; 4 x 1.5mL of 10 Optimized Buffer; 6mL
Code : A0
Additional Details : Weight : 1.51950kg
Product Code. 10550530