missing translation for 'onlineSavingsMsg'
Learn More

DyNAmo Flash DyNAmo Flash Probe qPCR Kit

Perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput.

Brand:  Thermo Scientific™ F455L

Code : 87

Additional Details : Weight : 0.00710kg

 View more versions of this product

Product Code. 10659599

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: DyNAmo Flash qPCR Mix (2X); 50X ROX dye, 25μM

Description

Description

DyNAmo Flash Probe qPCR Kit is the perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput. Significant time savings are achieved with both conventional and fast real-time PCR instruments. Performance of the DyNAmo Flash Probe qPCR Kit is based on an efficient hot start Thermus brockianus DNA polymerase. Sensitivity and the excellent amplification efficiency across a wide range of template concentrations guarantee reproducible results.

DyNAmo Flash Probe qPCR Kit is optimized for hydrolysis probes (e.g., TaqMan probes) and it can be used with both block- and capillary-based realtime PCR instruments.

  • Extremely fast protocols: combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • Included dUTP allows the use of UNG for prevention of carry-over contamination
  • Convenient 2X master mix and optimized protocols
Specifications

Specifications

Contains:
• 5 x 1 mL DyNAmo Flash qPCR Mix (2X)
• 1 x 250 μL 50X ROX dye, 25μM

Store at -20°C
Tube
qPCR
Fast
DyNAmo Flash Probe qPCR Kit
Real Time PCR SYBR Master Mix
Gene Expression
500 Reactions
5 x 1 mL
Primer-probe
High
Tbr DNA polymerase
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, Eppendorf™ MasterCycler™ RealPlex, QuantStudio™, ViiA™ 7 System, MJ Chromo4, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, BioRad iQ5, BioRad iCycler iQ, Corbett RotorGene, Cepheid SmartCycler, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, StepOne™ System, MJ Opticon, Stratagene Mx3000P
DyNAmo Flash
500 Reactions of 20 μL, 200 reactions of 50 μL
2X (DyNAmo Flash qPCR Mix), 50X (ROX Dye)
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.